Proteintech
Manufacturer Code:125151AP150UL
Catalog # 50-172-8036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NUFIP1 Fusion Protein Ag3197 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: nuclear FMRP interacting protein; Nuclear FMRP-interacting protein 1; nuclear fragile X mental retardation protein interacting protein 1; Nuclear fragile X mental retardation-interacting protein 1; NUFIP NUFIP1
Gene Aliases: bA540M5.1; NUFIP; NUFIP1
UniProt ID: (Human) Q9UHK0, (Mouse) Q9QXX8, (Rat) Q641W3
Entrez Gene ID: (Human) 26747, (Mouse) 27275, (Rat) 364430
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.