Proteintech
Manufacturer Code:157311AP150UL
Catalog # 50-173-0438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NUF2 Fusion Protein Ag8479 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer/testis antigen 106; CDCA1 hNuf2 hNuf2R hsNuf2 Kinetochore protein Nuf2 NUF2 NUF2R; cell division cycle associated 1; Cell division cycle-associated protein 1; hNuf2; hNuf2R; hsNuf2; Kinetochore protein Nuf2; NUF2, NDC80 kinetochore complex component, homolog
Gene Aliases: 2410003C07Rik; AU044112; C85691; CDCA1; CT106; NUF2; NUF2R
UniProt ID: (Human) Q9BZD4, (Rat) Q6AYL9, (Mouse) Q99P69
Entrez Gene ID: (Human) 83540, (Rat) 304951, (Mouse) 66977
Molecular Function:
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.