Novus Biologicals
Manufacturer Code:NBP273126FR
Catalog # NBP273126FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7A12 |
Immunogen | Full length recombinant protein of human NUDT9 (NP_076952) produced in HEK293T cells. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Adenosine diphosphoribose pyrophosphatase; ADP-ribose diphosphatase; ADP-ribose diphosphatase ADP-ribose phosphohydrolase ADP-ribose pyrosphosphatase NUDT9 ADPR-PPase EC 3.6.1.13 MGC3037 mitochondrial nucleoside diphosphate linked moiety X-type motif 9 nudix (nucleoside diphosphate linked moiety X)-type motif 9 Nudix motif 9 NUDT10; ADP-ribose phosphohydrolase; ADP-ribose pyrophosphatase, mitochondrial; ADP-ribose pyrosphosphatase NUDT9; ADPR-PPase; nucleoside diphosphate linked moiety X-type motif 9; Nucleoside diphosphate-linked moiety X motif 9; nudix (nucleoside diphosphate linked moiety X)-type motif 9; Nudix motif 9
Gene Aliases: NUDT10; NUDT9; PSEC0099; UNQ3012/PRO9771
UniProt ID: (Human) Q9BW91
Entrez Gene ID: (Human) 53343
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.