Invitrogen
Manufacturer Code:PA541085
Catalog # PIPA541085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human NUDT9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adenosine diphosphoribose pyrophosphatase; ADP-ribose diphosphatase; ADP-ribose phosphohydrolase; ADP-ribose pyrophosphatase; ADP-ribose pyrophosphatase, mitochondrial; ADP-ribose pyrosphosphatase NUDT9; ADPR-PPase; mitochondrial ADP-ribose pyrophosphatase-like protein; nucleoside diphosphate linked moiety X-type motif 9; Nucleoside diphosphate-linked moiety X motif 9; nudix (nucleoside diphosphate linked moiety X)-type motif 9; Nudix (nucleoside diphosphate linked moiety X)-type motif 9 NUDT10; nudix -type motif 9; Nudix motif 9; nudix-type motif 9
Gene Aliases: 1190002C07Rik; ADPR-PPase; AI462474; fu33g07; NUDT10; NUDT9; PSEC0099; sb:cb392; UNQ3012/PRO9771; wu:fj16b09; wu:fu33g07; zgc:63924
UniProt ID: (Human) Q9BW91, (Rat) Q5XIG0, (Mouse) Q8BVU5
Entrez Gene ID: (Bovine) 517589, (Pig) 100521385, (Sheep) 100529154, (Human) 53343, (Rabbit) 100347918, (Dog) 478469, (Horse) 100052874, (Rat) 305149, (Guinea pig) 100716076, (Mouse) 74167, (Zebrafish) 406661
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.