Abnova
Manufacturer Code:PAB18875
Catalog # 89-120-436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids 2-236 of human NUDT7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant NUDT7.
For Research Use Only
Protein Aliases: Nucleoside diphosphate-linked moiety X motif 7; nudix (nucleoside diphosphate linked moiety X)-type motif 7; Nudix motif 7; Peroxisomal coenzyme A diphosphatase NUDT7
Gene Aliases: NUDT7
UniProt ID: (Human) P0C024
Entrez Gene ID: (Human) 283927
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.