OriGene
Manufacturer Code:CF502330
Catalog # 50-166-1241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9F5 |
Immunogen | Full length human recombit protein of human NUDT6 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Antisense basic fibroblast growth factor; ASFGF2 FGF-AS FGF2AS GFG-1 GFG1; Nucleoside diphosphate-linked moiety X motif 6; nudix (nucleoside diphosphate linked moiety X)-type motif 6; Nudix motif 6; Protein GFG
Gene Aliases: ASFGF2; FGF-AS; FGF2AS; GFG-1; GFG1; NUDT6
UniProt ID: (Human) P53370
Entrez Gene ID: (Human) 11162
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.