Invitrogen
Manufacturer Code:PA5112142
Catalog # PIPA5112142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human ADP-sugar Pyrophosphatase/NUDT5 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 8-oxo-dGDP phosphatase; 8-oxo-dGDP phosphatase ADP-sugar pyrophosphatase HSPC115 Nuclear ATP-synthesis protein NUDIX5 Nucleoside diphosphate-linked moiety X motif 5 nudix (nucleoside diphosphate linked moiety X)-type motif 5 nudix hydrolase 5 Nudix motif 5 NUDIX5 nudix-type motif 5 NUDT5 YSA1 YSA1H YSAH1; ADP-sugar pyrophosphatase; Nuclear ATP-synthesis protein NUDIX5; Nucleoside diphosphate-linked moiety X motif 5; nudix (nucleoside diphosphate linked moiety X)-type motif 5; Nudix motif 5; YSA1H
Gene Aliases: HSPC115; NUDIX5; NUDT5; YSA1; YSA1H; YSAH1
UniProt ID: (Human) Q9UKK9
Entrez Gene ID: (Human) 11164
Molecular Function: hydrolase pyrophosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.