OriGene
Manufacturer Code:CF503828
Catalog # 50-166-2415
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B4 |
Immunogen | Full length human recombit protein of human NUDT18 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2-hydroxy-dADP phosphatase; 7,8-dihydro-8-oxoguanine phosphatase; 8-oxo-dGDP phosphatase NUDT18; MTH3; MutT homolog 3; mutT human homolog 3; Nucleoside diphosphate-linked moiety X motif 18; nudix (nucleoside diphosphate linked moiety X)-type motif 18; Nudix motif 18
Gene Aliases: MTH3; NUDT18
UniProt ID: (Human) Q6ZVK8
Entrez Gene ID: (Human) 79873
Molecular Function: hydrolase phosphorylase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.