Novus Biologicals
Manufacturer Code:NBP29442220UL
Catalog # NB023395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 60-130 of human NUDT16 (NP_689608.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.6.1 EC 3.6.1.- FLJ31265 FLJ34034 FLJ36248 Nucleoside diphosphate-linked moiety X motif 16 nudix (nucleoside diphosphate linked moiety X)-type motif 16 Nudix motif 16 U8 snoRNA-binding protein H29K U8 snoRNA-decapping enzyme; IDP phosphatase; IDPase; Inosine diphosphate phosphatase; m7GpppN-mRNA hydrolase; Nucleoside diphosphate-linked moiety X motif 16; nudix (nucleoside diphosphate linked moiety X)-type motif 16; Nudix hydrolase 16; Nudix motif 16; testicular tissue protein Li 129; U8 snoRNA-binding protein H29K; U8 snoRNA-decapping enzyme
Gene Aliases: NUDT16
UniProt ID: (Human) Q96DE0
Entrez Gene ID: (Human) 131870
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.