Proteintech
Manufacturer Code:128891AP150UL
Catalog # 50-172-8325
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NUDT16 Fusion Protein Ag3925 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IDP phosphatase; IDPase; Inosine diphosphate phosphatase; m7GpppN-mRNA hydrolase; Nucleoside diphosphate-linked moiety X motif 16; nudix (nucleoside diphosphate linked moiety X)-type motif 16; Nudix hydrolase 16; Nudix motif 16; Nudix motif 16 NUDT16 U8 snoRNA binding protein H29K U8 snoRNA decapping enzyme; testicular tissue protein Li 129; U8 snoRNA-binding protein H29K; U8 snoRNA-decapping enzyme
Gene Aliases: NUDT16
UniProt ID: (Human) Q96DE0
Entrez Gene ID: (Human) 131870
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.