OriGene
Manufacturer Code:CF811514
Catalog # 50-167-0701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI11F3 |
Immunogen | Full length human recombit protein of human NUDT15 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 8-oxo-dGTPase NUDT15; MTH2; MTH2 NUDT15D; MutT homolog 2; Nucleoside diphosphate-linked moiety X motif 15; Nucleoside diphosphate-linked to another moiety X hydrolase 15; Nucleotide triphosphate diphosphatase NUDT15; nudix (nucleoside diphosphate linked moiety X)-type motif 15; Nudix hydrolase 15; Nudix motif 15; probable 7,8-dihydro-8-oxoguanine triphosphatase NUDT15
Gene Aliases: MTH2; NUDT15; NUDT15D
UniProt ID: (Human) Q9NV35
Entrez Gene ID: (Human) 55270
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.