Invitrogen
Manufacturer Code:PA543258
Catalog # PIPA543258
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human NUDT12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DeNADding enzyme NUDT12; NAD-capped RNA hydrolase NUDT12; NADH pyrophosphatase NUDT12; nucleoside diphosphate linked moiety X-type motif 12; Nucleoside diphosphate-linked moiety X motif 12; nudix (nucleoside diphosphate linked moiety X)-type motif 12; Nudix (nucleoside diphosphate linked moiety X)-type motif 12 DKFZP761I172; nudix (nucleoside diphosphnudix (nucleoside diphosphate linked moiety X)-type motif 12ate linked moiety X)-type motif 12; Nudix motif 12; nudix-type motif 12; peroxisomal NADH pyrophosphatase NUDT12
Gene Aliases: 0610016O18Rik; NUDT12
UniProt ID: (Human) Q9BQG2, (Bovine) Q29RH3, (Mouse) Q9DCN1
Entrez Gene ID: (Pig) 100512459, (Rabbit) 100354787, (Dog) 488881, (Horse) 100073208, (Human) 83594, (Bovine) 617720, (Rat) 367323, (Mouse) 67993, (Guinea pig) 100735803
Molecular Function:
hydrolase
phosphorylase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.