Novus Biologicals
Manufacturer Code:NBP256969
Catalog # NBP256969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TASKASKEKTPSPKEEDEEPESPPEKKTSTSPPPEKSGDEGSEDEAPSGED |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ21480 FLJ32016 FLJ38536 NUCKSnuclear ubiquitous casein and cyclin-dependent kinases substrate nuclear casein kinase and cyclin-dependent kinase substrate 1 nuclear ubiquitous casein kinase and cyclin-dependent kinase substrate P1 potential LAG1 interactor; nuclear casein kinase and cyclin-dependent kinase substrate 1; Nuclear ubiquitous casein and cyclin-dependent kinase substrate 1; nuclear ubiquitous casein kinase and cyclin-dependent kinase substrate; P1; potential LAG1 interactor
Gene Aliases: JC7; NUCKS; NUCKS1
UniProt ID: (Human) Q9H1E3
Entrez Gene ID: (Human) 64710
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.