Proteintech
Manufacturer Code:267121AP150UL
Catalog # 50-173-5433
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NUCB2/nesfatin-1 Fusion Protein Ag25152 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA binding protein NEFA Gastric cancer antigen Zg4 NEFA NUCB2 nucleobindin 2; DNA-binding protein NEFA; Epididymis secretory protein Li 109; Gastric cancer antigen Zg4; nesfatin 1; Nesfatin-1; Nucleobindin-2; nucleobinding 2; prepronefastin; Prepronesfatin
Gene Aliases: AI607786; Calnuc; HEL-S-109; NEFA; Nesfatin-1; NUCB2; p54
UniProt ID: (Human) P80303, (Mouse) P81117, (Rat) Q9JI85
Entrez Gene ID: (Human) 4925, (Mouse) 53322, (Rat) 59295
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.