Invitrogen
Manufacturer Code:PA579767
Catalog # PIPA579767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse Nucleobindin 2 recombit protein (Position: V25-L106). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-binding protein NEFA; Nesfatin-1; Nucleobindin-2; Nucleobindin-2 DNA-binding protein NEFA Prepronesfatin Nesfatin-1 Nucb2 Nefa; prepronefastin; Prepronesfatin
Gene Aliases: AI607786; Calnuc; Nefa; Nesfatin-1; Nucb2; p54
UniProt ID: (Mouse) P81117, (Rat) Q9JI85
Entrez Gene ID: (Mouse) 53322, (Rat) 59295
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.