Invitrogen
Manufacturer Code:PA577384
Catalog # PIPA577384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)DKTKVHNVEPVESAR corresponding to amino acid residues 30-44 of rat Nucleobindin-2/Nesfatin-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA-binding protein NEFA; Epididymis secretory protein Li 109; Gastric cancer antigen Zg4; nesfatin 1; Nesfatin-1; Nucleobindin-2; Nucleobindin-2/Nesfatin-1; nucleobinding 2; prepronefastin; Prepronesfatin
Gene Aliases: AI607786; Calnuc; HEL-S-109; NEFA; Nesfatin-1; NUCB2; p54
UniProt ID: (Human) P80303, (Mouse) P81117, (Rat) Q9JI85
Entrez Gene ID: (Human) 4925, (Mouse) 53322, (Rat) 59295
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.