Abnova
Manufacturer Code:PAB18034
Catalog # 89-119-620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus amino acids of human NUCB2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NUCB2.
For Research Use Only
Protein Aliases: DNA-binding protein NEFA; Epididymis secretory protein Li 109; Gastric cancer antigen Zg4; NEFA; nesfatin 1; Nesfatin-1; Nucleobindin-2; nucleobinding 2; prepronefastin; Prepronesfatin
Gene Aliases: HEL-S-109; NEFA; NUCB2
UniProt ID: (Human) P80303
Entrez Gene ID: (Human) 4925
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.