Novus Biologicals
Manufacturer Code:NBP294547100UL
Catalog # NB023618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 142-271 of human NUBP2 (NP_036357.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CFD1C447E6.1 (nucleotide binding protein 1 (E.coli MinD like) ) cytosolic Fe-S cluster assembly factor NUBP2 homolog of yeast cytosolic Fe-S cluster deficient 1 NBP 2 NUBP1 nucleotide binding protein 2 (E.coli MinD like) nucleotide binding protein 2 (MinD homolog E. coli) Nucleotide-binding protein 2; Cytosolic Fe-S cluster assembly factor NUBP2; homolog of yeast cytosolic Fe-S cluster deficient 1; NBP 2; nucleotide binding protein 2 (MinD homolog, E. coli); Nucleotide-binding protein 2
Gene Aliases: CFD1; NBP 2; NUBP1; NUBP2
UniProt ID: (Human) Q9Y5Y2
Entrez Gene ID: (Human) 10101
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.