Invitrogen
Manufacturer Code:MA537545
Catalog # PIMA537545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 378CT15.4.5 |
Immunogen | purified His-tagged NUAK2 protein (fragment) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 1200013b22rik EMBL:AAH81899.1} mKIAA0537 NUAK family kinase 2 NUAK family SNF1-like kinase 2 NUAK family SNF1-like kinase 2 NUAK2 nuak2 {ECO:0000312 Omphalocele kinase 2 OMPHK2 SNARK SNF1/AMP activated protein kinase SNF1/AMP kinase-related kinase SNF1/AMP-activated protein kinase UV126; NUAK family SNF1-like kinase 2; NUAK family, SNF1-like kinase, 2; NUAK2; Omphalocele kinase 2; SNARK; SNF1/AMP activated protein kinase; SNF1/AMP kinase-related kinase
Gene Aliases: NUAK2; OMPHK2; SNARK
UniProt ID: (Human) Q9H093
Entrez Gene ID: (Human) 81788
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.