Invitrogen
Manufacturer Code:PA513959
Catalog # PIPA513959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 67-96 amino acids from the N-terminal region of human NUAK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NUAK family SNF1-like kinase 2; NUAK family SNF1-like kinase 2 SNF1/AMP kinase-related kinase SNARK NUAK2; NUAK family, SNF1-like kinase, 2; NUAK2; Omphalocele kinase 2; SNARK; SNF1/AMP activated protein kinase; SNF1/AMP kinase-related kinase
Gene Aliases: NUAK2; OMPHK2; SNARK
UniProt ID: (Human) Q9H093
Entrez Gene ID: (Human) 81788
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.