Invitrogen
Manufacturer Code:PA5102344
Catalog # PIPA5102344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NUAK2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1200013b22rik EMBL:AAH81899.1} mKIAA0537 NUAK family kinase 2 NUAK family SNF1-like kinase 2 NUAK family SNF1-like kinase 2 NUAK2 nuak2 {ECO:0000312 Omphalocele kinase 2 OMPHK2 SNARK SNF1/AMP activated protein kinase SNF1/AMP kinase-related kinase SNF1/AMP-activated protein kinase UV126; NUAK family SNF1-like kinase 2; NUAK family, SNF1-like kinase, 2; NUAK2; nuak2 {ECO:0000312|EMBL:AAH81899.1}; Omphalocele kinase 2; SNARK; SNF1/AMP activated protein kinase; SNF1/AMP kinase-related kinase; SNF1/AMP-activated protein kinase
Gene Aliases: 1200013B22Rik; mKIAA0537; NUAK2; OMPHK2; SNARK; UV126
UniProt ID: (Human) Q9H093, (Mouse) Q8BZN4, (Rat) Q66HE5
Entrez Gene ID: (Human) 81788, (Mouse) 74137, (Rat) 289419
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.