Invitrogen
Manufacturer Code:PA141575
Catalog # 01-675-194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 150-200 of human Neurotensin receptor 2 . |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Levocabastine-sensitive neurotensin receptor; low affinity neurotensin receptor; Low-affinity levocabastine-sensitive neurotensin receptor; Neurotensin receptor type 2; neurotensin receptor, type 2; NT-R-2; NTR2; NTRL
Gene Aliases: NT2R; NTR2; NTRL; NTSR2
UniProt ID: (Human) O95665, (Mouse) P70310
Entrez Gene ID: (Human) 23620, (Mouse) 18217
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.