Invitrogen
Manufacturer Code:PA534028
Catalog # PIPA534028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human NTSR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: High-affinity levocabastine-insensitive neurotensin receptor; Neurotensin receptor type 1; NT-R-1; NTR1; NTRH; NTSR1 Neurotensin receptor type 1 NT-1r NT1 receptor NTR NTS1 Neurotensin receptor 1 NT-R-1 NTR1 NTRH NTRR Neurotensin receptor ntr1 NTS-1 Neurotensin type 1 receptor NT receptor-1 RNTR1
Gene Aliases: NTR; NTRR; NTSR1
UniProt ID: (Human) P30989
Entrez Gene ID: (Human) 4923
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.