Abnova
Manufacturer Code:PAB14331
Catalog # 89-117-005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Glycosylated mouse extracellular domain Ntrk2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with deionized water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial Ntrk2.
For Research Use Only
Protein Aliases: AI848316; BDNF/NT-3 growth factors receptor; C030027L06Rik; GP145-TrkB/GP95-TrkB; Neurotrophic tyrosine kinase receptor type 2; neurotrophic tyrosine receptor kinase type 2; Tkrb; Trk-B; trkB; TrkB tyrosine kinase
Gene Aliases: GP145-TrkB/GP95-TrkB; Ntrk2; Tkrb; trk-B; Trkb
UniProt ID: (Mouse) P15209
Entrez Gene ID: (Mouse) 18212
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.