Novus Biologicals
Manufacturer Code:NBP299976FR
Catalog # NB090507
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant NTNG1 (NP_055732.2 Met1-Gly408). |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: axon guidance molecule; axon guidance molecule KIAA0976YLSR571 laminet 1 laminet-1 Lmnt1 netrin G1 netrin G1f netrin-G1; laminet 1; Laminet-1; Netrin-G1
Gene Aliases: KIAA0976; LMNT1; NTNG1; UNQ571/PRO1133
UniProt ID: (Human) Q9Y2I2
Entrez Gene ID: (Human) 22854
Molecular Function: extracellular matrix linker protein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.