Novus Biologicals
Manufacturer Code:NBP211809AF488
Catalog # NB211809488
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0482-9H27 |
Immunogen | Human NTN4 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Beta-netrin; beta-netrin FLJ23180 hepar-derived netrin-like protein netrin 4 PRO3091; Hepar-derived netrin-like protein; Netrin-4
Gene Aliases: NTN4; PRO3091
UniProt ID: (Human) Q9HB63
Entrez Gene ID: (Human) 59277
Molecular Function: extracellular matrix linker protein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.