Invitrogen
Manufacturer Code:PA547077
Catalog # PIPA547077
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Netrin-4 Leu19-Val628 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-netrin; beta-Netrin NTN4 CDY CDY1A chromodomain protein Y chromosome 1 chromodomain protein Y-linked 1 FLJ77185 testis-specific chromodomain protein on Y testis-specific chromodomain protein Y 1; Netrin-4
Gene Aliases: Ntn4
UniProt ID: (Mouse) Q9JI33
Entrez Gene ID: (Mouse) 57764
Molecular Function: extracellular matrix linker protein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.