Invitrogen
Manufacturer Code:PA577131
Catalog # PIPA577131
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human NTMT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha N-terminal protein methyltransferase 1A; methyltransferase like 11A; Methyltransferase-like protein 11A; N-terminal RCC1 methyltransferase; N-terminal Xaa-Pro-Lys N-methyltransferase 1; N-terminal Xaa-Pro-Lys N-methyltransferase 1 Alpha N-terminal protein methyltransferase 1A Methyltransferase-like protein 11A N-terminal RCC1 methyltransferase X-Pro-Lys N-terminal protein methyltransferase 1A NTM1A NTMT1 C9orf32 METTL11A NRMT NRMT1 AD-003; N-terminal Xaa-Pro-Lys N-methyltransferase 1, N-terminally processed; NTM1A; X-Pro-Lys N-terminal protein methyltransferase 1A
Gene Aliases: 2610205E22Rik; AD-003; AL033331; AL033332; C9orf32; HOMT1A; METTL11A; NRMT; NRMT1; NTM1A; NTMT1
UniProt ID: (Human) Q5SZB9, (Mouse) Q8R2U4
Entrez Gene ID: (Human) 28989, (Mouse) 66617
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.