Invitrogen
Manufacturer Code:PA5111469
Catalog # PIPA5111469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human NTMT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610205E22Rik AD-003 AL033331 AL033332 Alpha N-terminal protein methyltransferase 1A C9orf32 HOMT1A Methyltransferase-like protein 11A METTL11A NRMT NRMT1 N-terminal RCC1 methyltransferase N-terminal Xaa-Pro-Lys N-methyltransferase 1 N-terminal Xaa-Pro-Lys N-methyltransferase 1 N-terminally processed NTM1A NTMT1 RGD1306582 X-Pro-Lys N-terminal protein methyltransferase 1A; Alpha N-terminal protein methyltransferase 1A; Methyltransferase-like protein 11A; N-terminal RCC1 methyltransferase; N-terminal Xaa-Pro-Lys N-methyltransferase 1; N-terminal Xaa-Pro-Lys N-methyltransferase 1, N-terminally processed; NTM1A; X-Pro-Lys N-terminal protein methyltransferase 1A
Gene Aliases: AD-003; C9orf32; HOMT1A; METTL11A; NRMT; NRMT1; NTM1A; NTMT1
UniProt ID: (Human) Q5SZB9
Entrez Gene ID: (Human) 28989
Molecular Function: methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.