Invitrogen
Manufacturer Code:PA539305
Catalog # PIPA539305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human NT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hNT; IgLON family member 2; IGLON2 NT; Neurotrimin
Gene Aliases: HNT; IGLON2; NT; NTM; NTRI; UNQ297/PRO337
UniProt ID: (Human) Q9P121
Entrez Gene ID: (Human) 50863
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.