Invitrogen
Manufacturer Code:PA5112631
Catalog # PIPA5112631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Endonuclease III-like protein 1 protein (31-312AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bifunctional DNA N-glycoslyase/DNA-(apurinic or apyrimidinic site) lyase; bifunctional DNA N-glycoslyase/DNA-(apurinic or apyrimidinic site) lyase Bifunctional DNA N-glycosylase/DNA-(apurinic or apyrimidinic site) lyase DNA glycoslyase/AP lyase DNA glycosylase/AP lyase Endonuclease III-like protein 1 FAP3 hNTH1 nth (endonuclease III)-like 1 (E.coli) nth endonuclease III-like 1 NTH1 NTHL 1 NTHL1 nth-like DNA glycosylase 1 OCTS 3 OCTS3 thymine glycol DNA glycosylase/AP lyase; Bifunctional DNA N-glycosylase/DNA-(apurinic or apyrimidinic site) lyase; DNA glycoslyase/AP lyase; DNA glycosylase/AP lyase; Endonuclease III-like protein 1; hNTH1; nth endonuclease III-like 1; thymine glycol DNA glycosylase/AP lyase
Gene Aliases: FAP3; hNTH1; NTH1; NTHL1; OCTS3
UniProt ID: (Human) P78549, (Mouse) O35980
Entrez Gene ID: (Human) 4913, (Mouse) 18207
Molecular Function: DNA binding protein DNA glycosylase endodeoxyribonuclease glycosidase hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.