Proteintech
Manufacturer Code:149181AP150UL
Catalog # 50-172-9834
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NTHL1 Fusion Protein Ag6734 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bifunctional DNA N-glycoslyase/DNA-(apurinic or apyrimidinic site) lyase; Bifunctional DNA N-glycosylase/DNA-(apurinic or apyrimidinic site) lyase; DNA glycoslyase/AP lyase; DNA glycosylase/AP lyase; Endonuclease III-like protein 1; hNTH1; nth endonuclease III-like 1; NTH1 NTHL1 OCTS3; thymine glycol DNA glycosylase/AP lyase
Gene Aliases: FAP3; hNTH1; NTH1; NTHL1; OCTS3
UniProt ID: (Human) P78549, (Mouse) O35980
Entrez Gene ID: (Human) 4913, (Mouse) 18207, (Rat) 29541
Molecular Function:
DNA binding protein
DNA glycosylase
endodeoxyribonuclease
glycosidase
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.