Invitrogen
Manufacturer Code:PA5111291
Catalog # PIPA5111291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human NTF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900040K06Rik AI462899 GLC10 GLC1O neurotrophin 4 Neurotrophin 4/5 neurotrophin 5 Neurotrophin-4 Neurotrophin-5 Neutrophic factor 4 Neutrophic factor 5 Nt4 NT-4 NT4/5 NT-4/5 NT4/NT5 NT4P NT5 NT-5 Ntf4 Ntf5 Ntf-5; neurotrophic factor 4; neurotrophic factor 5; neurotrophin 5 (neurotrophin 4/5); Neurotrophin-4; Neurotrophin-5; Neutrophic factor 4; NT-4; NT-5
Gene Aliases: GLC10; GLC1O; NT-4; NT-4/5; NT-5; NT4; NT5; NTF4; NTF5
UniProt ID: (Human) P34130
Entrez Gene ID: (Human) 4909
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.