Invitrogen
Manufacturer Code:PA5102705
Catalog # PIPA5102705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NTF4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900040K06Rik AI462899 GLC10 GLC1O neurotrophic factor 4 neurotrophic factor 5 neurotrophin 4 Neurotrophin 4/5 neurotrophin 5 Neurotrophin 5 (neurotrophin 4/5) neurotrophin 5 (neurotrophin 4/5) neurotrophin-4 Neurotrophin-5 neutrophic factor 4 Neutrophic factor 5 Nt4 NT-4 NT4/5 NT-4/5 NT4/NT5 NT4P NT5 NT-5 Ntf4 Ntf5 Ntf-5; neurotrophic factor 4; neurotrophic factor 5; Neurotrophin 5 (neurotrophin 4/5); neurotrophin 5 (neurotrophin 4/5); Neurotrophin-4; Neurotrophin-5; Neutrophic factor 4; NT-4; NT-5
Gene Aliases: 2900040K06Rik; AI462899; GLC10; GLC1O; NT-4; NT-4/5; NT-5; NT4; NT4/5; NT4P; NT5; Ntf-5; NTF4; NTF5
UniProt ID: (Human) P34130, (Rat) P34131, (Mouse) Q80VU4
Entrez Gene ID: (Human) 4909, (Rat) 25730, (Mouse) 78405
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.