Invitrogen
Manufacturer Code:MA524883
Catalog # PIMA524883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5A2 |
Immunogen | Recombit protein expressed in E.coli corresponding to amino acids 139-257 of NT3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HDNF; Nerve growth factor 2; Neurotrophic factor; Neurotrophin-3; NGF-2; NT-3; NTF3 HDNF NGF2 Nerve growth factor 2 NGF-2 NT-3 Neurotrophic factor Neurotrophin 3 Neurotrophin-3
Gene Aliases: HDNF; NGF-2; NGF2; NT-3; NT3; NTF3
UniProt ID: (Human) P20783
Entrez Gene ID: (Human) 4908
Molecular Function:
growth factor
neurotrophic factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.