Invitrogen
Manufacturer Code:PA514861
Catalog # PIPA514861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 152-184 amino acids from the C-terminal region of human NTF3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HDNF; Nerve growth factor 2; Neurotrophic factor; Neurotrophin-3; Neurotrophin-3 &lsqbPrecursor] NT-3 Neurotrophic factor HDNF Nerve growth factor 2 NGF-2 NTF3; NGF-2; NT-3
Gene Aliases: AI316846; AI835689; HDNF; NGF-2; NGF2; NT-3; NT3; Ntf-3; NTF3
UniProt ID: (Human) P20783, (Mouse) P20181
Entrez Gene ID: (Human) 4908, (Mouse) 18205
Molecular Function:
growth factor
neurotrophic factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.