Invitrogen
Manufacturer Code:PA118385
Catalog # 01-670-637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Inhibition Assays (IA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues Y(139) A E H K S H R G E Y(149) of human mouse and rat NT3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HDNF; Nerve growth factor 2; Neurotrophic factor; Neurotrophin-3; neurotrophin-3 (HDNF/NT-3); Neurotrophin-3 Neurotrophic factor HDNF Nerve growth factor 2 NGF-2 Ntf3 Ntf-3; NGF-2; NT-3
Gene Aliases: HDNF; neurotrophin-3; NGF-2; NGF2; NT-3; NT3; Ntf-3; NTF3
UniProt ID: (Chicken) P25433, (Human) P20783, (Rat) P18280
Entrez Gene ID: (Chicken) 428099, (Human) 4908, (Rat) 81737
Molecular Function: growth factor neurotrophic factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.