Invitrogen
Manufacturer Code:PA535939
Catalog # PIPA535939
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1017 and 1270 of NTE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuropathy target esterase; patatin-like phospholipase domain containing 6; patatin-like phospholipase domain containing 6 NTEMND SPG39 PNPLA6 iPLA2delta; Patatin-like phospholipase domain-containing protein 6; Swiss cheese; Swiss-cheese
Gene Aliases: AI661849; MSws; Nte; Pnpla6; RGD1564611
UniProt ID: (Mouse) Q3TRM4
Entrez Gene ID: (Mouse) 50767, (Rat) 360753
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.