Invitrogen
Manufacturer Code:PA5101720
Catalog # PIPA5101720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PNPLA6 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI661849 BNHS CD73 iPLA2delta LNMS MSws Neuropathy target esterase NTE NTEMND OMCS patatin like phospholipase domain containing 6 patatin-like phospholipase domain containing 6 patatin-like phospholipase domain-containing protein 6 PNPLA6 RGD1564611 SPG39 Swiss cheese Swiss-cheese sws; Neuropathy target esterase; patatin-like phospholipase domain containing 6; Patatin-like phospholipase domain-containing protein 6; Swiss cheese; Swiss-cheese
Gene Aliases: AI661849; BNHS; iPLA2delta; LNMS; MSws; NTE; NTEMND; OMCS; PNPLA6; SPG39; sws
UniProt ID: (Human) Q8IY17, (Mouse) Q3TRM4
Entrez Gene ID: (Human) 10908, (Mouse) 50767
Molecular Function: esterase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.