Invitrogen
Manufacturer Code:PA588724
Catalog # PIPA588724
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human SLC10A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bile acid cotransporting polypeptide; Na(+)/bile acid cotransporter; Na(+)/taurocholate transport protein; sodium bile acid cotransporting polypeptide; sodium-taurocholate cotransporting polypeptide; Sodium/bile acid cotransporter; Sodium/taurocholate cotransporting polypeptide; Solute carrier family 10 member 1
Gene Aliases: Ntcp; Slc10a1
UniProt ID: (Mouse) O08705
Entrez Gene ID: (Mouse) 20493
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.