Invitrogen
Manufacturer Code:PA580000
Catalog # PIPA580000
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human SLC10A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bile acid cotransporting polypeptide; Cell growth-inhibiting gene 29 protein; growth-inhibiting protein 29; Na(+)/bile acid cotransporter; Na(+)/taurocholate transport protein; NA-dependent cholate transporting protein; Na/taurocholate cotransporting polypeptide; sodium bile acid cotransporting polypeptide; sodium-dependent bile acid cotransporter; sodium-dependent taurocholate cotransporting polypeptide; sodium-taurocholate cotransporting polypeptide; Sodium/bile acid cotransporter; Sodium/bile acid cotransporter Cell growth-inhibiting gene 29 protein Na(+)/bile acid cotransporter Na(+)/taurocholate transport protein Sodium/taurocholate cotransporting polypeptide Solute carrier family 10 member 1 SLC10A1 NTCP GIG29; sodium/taurocholate cotransporter; Sodium/taurocholate cotransporting polypeptide; solute carrier family 10 (sodium/bile acid cotransporter family), member 1; solute carrier family 10 (sodium/bile acid cotransporter), member 1; Solute carrier family 10 member 1; solute carrier family 10, member 1
Gene Aliases: GIG29; NTCP; Ntcp1; SBACT; SLC10A1
UniProt ID: (Human) Q14973, (Mouse) O08705, (Rat) P26435
Entrez Gene ID: (Human) 6554, (Mouse) 20493, (Rat) 24777
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.