Novus Biologicals
Manufacturer Code:NBP298491G
Catalog # NB104320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human NTAN1 fragment. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp666E058 EC 3.5.1.- N-terminal asparagine amidase N-terminal asparagine amidohydrolase PNAA PNAD Protein NH2-terminal asparagine amidohydrolase Protein NH2-terminal asparagine deamidase Protein N-terminal Asn amidase protein N-terminal asparagine amidohydrolase Protein NTN-amidase; PNAA; PNAD; protein N-terminal Asn amidase; protein N-terminal asparagine amidase; Protein N-terminal asparagine amidohydrolase; Protein NH2-terminal asparagine amidohydrolase; Protein NH2-terminal asparagine deamidase; protein NTN-amidase
Gene Aliases: NTAN1; PNAA; PNAD
UniProt ID: (Human) Q96AB6
Entrez Gene ID: (Human) 123803
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.