Novus Biologicals
Manufacturer Code:NBP15468420UL
Catalog # NBP15468420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to NT5M(5'3'-nucleotidase mitochondrial) The peptide sequence was selected from the middle region of NT5M. Peptide sequence KYAWVEKYFGPDFLEQIVLTRDKTVVSADLLIDDRPDITGAEPTPSWEHV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3'-nucleotidase mitochondrialDNT2 5' 5' nucleotidase mitochondrial 5(3)-deoxyribonucleotidase Deoxy-5'-nucleotidase 2 dNT2 dNT-25'(3')-deoxyribonucleotidase mitochondrial EC 3.1.3 EC 3.1.3.- mdN mitochondrial 5' nucleotidase; 5' nucleotidase, mitochondrial; 5'(3')-deoxyribonucleotidase, mitochondrial; 5',3'-nucleotidase, mitochondrial; 5(3)-deoxyribonucleotidase; Deoxy-5'-nucleotidase 2; dNT-2; mitochondrial 5' nucleotidase
Gene Aliases: dNT-2; DNT2; mdN; NT5M
UniProt ID: (Human) Q9NPB1
Entrez Gene ID: (Human) 56953
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.