Novus Biologicals
Manufacturer Code:NBP273078C
Catalog # NBP273078C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E11 |
Immunogen | Full length recombinant protein of human NT5DC1 (NP_689942) produced in HEK293T cells. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 5&prime-nucleotidase domain containing 15&prime-nucleotidase domain-containing protein 1 5&prime-nucleotidase cytosolic II-like 1 C6orf200 dJ486I3.1 LP2642 MGC131837 MGC243025&prime-nucleotidase cytosolic II-like 1 protein NT5C2L1; 5'-nucleotidase domain-containing protein 1; 5'-nucleotidase, cytosolic II-like 1 protein
Gene Aliases: C6orf200; LP2642; NT5C2L1; NT5DC1
UniProt ID: (Human) Q5TFE4
Entrez Gene ID: (Human) 221294
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.