Novus Biologicals
Manufacturer Code:NBP238818
Catalog # NBP238818
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: IVSNYMDFNEDGFLQGFKGQLIHTYNKNSSVCENCGYFQQLEGKTNVILLGDSIG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-nucleotidase cytosolic III-like; 5'-nucleotidase, cytosolic III-like; 7-methylguanosine nucleotidase; 7-methylguanosine phosphate-specific 5'-nucleotidase; cN-III-like protein; Cytosolic 5'-nucleotidase 3B; Cytosolic 5'-nucleotidase III-like protein; N(7)-methylguanylate 5'-phosphatase
Gene Aliases: cN-IIIB; NT5C3B; NT5C3L
UniProt ID: (Human) Q969T7
Entrez Gene ID: (Human) 115024
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.