Novus Biologicals
Manufacturer Code:NBP29333220UL
Catalog # NB028549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 37-336 of human NT5C3A (NP_001002010.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5&prime-nucleotidase cytosolic III cN-IIIMGC87828 cytosolic 5&prime-nucleotidase 3 Cytosolic 5&prime-nucleotidase III p36 P5N1 P5N-1 P5&primeN-1MGC27337 PN-IMGC87109 PSN1UMPH Pyrimidine 5&prime-nucleotidase 1 UMPH1EC 3.1.3.5 uridine 5&prime monophosphate hydrolase 1 Uridine 5&prime-monophosphate hydrolase 1; 5'-nucleotidase, cytosolic III; 7-methylguanosine nucleotidase; 7-methylguanosine phosphate-specific 5'-nucleotidase; cN-III; Cytosolic 5'-nucleotidase 3; Cytosolic 5'-nucleotidase 3A; Cytosolic 5'-nucleotidase III; lupin; p36; P5'N-1; Pyrimidine 5'-nucleotidase 1; Uridine 5'-monophosphate hydrolase 1
Gene Aliases: cN-III; HSPC233; hUMP1; NT5C3; NT5C3A; p36; P5'N-1; P5N-1; P5N1; PN-I; POMP; PSN1; UMPH; UMPH1
UniProt ID: (Human) Q9H0P0
Entrez Gene ID: (Human) 51251
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.