Proteintech
Manufacturer Code:113931AP150UL
Catalog # 50-172-7119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NT5C3 Fusion Protein Ag1949 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5&prime nucleotidase cytosolic III cN III Cytosolic 5 nucleotidase 3 Cytosolic 5 nucleotidase III NT5C3 p36 P5N 1 P5&primeN 1 P5N1 PN I PSN1 Pyrimidine 5 nucleotidase 1 UMPH UMPH1; 5'-nucleotidase, cytosolic III; 5'-nucleotidase, pyrimidine; 7-methylguanosine nucleotidase; 7-methylguanosine phosphate-specific 5'-nucleotidase; cN-III; Cytosolic 5'-nucleotidase 3; Cytosolic 5'-nucleotidase 3A; Cytosolic 5'-nucleotidase III; Lupin; p36; P5'N-1; P5N-1; Pyrimidine 5'-nucleotidase 1; Uridine 5'-monophosphate hydrolase 1; uridine monophosphatase 1
Gene Aliases: 1600024P05Rik; 2610206B05Rik; 3110004A18Rik; AI842776; cN-III; HSPC233; hUMP1; lupin; NT5C3; NT5C3A; Nt5y; p36; P5'N-1; P5N-1; P5N1; PN-1; PN-I; POMP; PSN1; UMPH; Umph-1; UMPH1
UniProt ID: (Human) Q9H0P0, (Mouse) Q9D020
Entrez Gene ID: (Human) 51251, (Mouse) 107569, (Rat) 312373
Molecular Function:
esterase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.