Invitrogen
Manufacturer Code:PA5103777
Catalog # PIPA5103777
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NT5C2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010002I23Rik 5'-nucleotidase (purine) cytosolic type B 5'-nucleotidase cytosolic II CnII cN-II Cytosolic 5'-nucleotidase II cytosolic purine 5'-nucleotidase ENSMUSG00000025039 Gm9751 GMP IMP-specific 5'-NT Nt5b NT5C2 NT5CP PNT5 RP11-30H12.3 SPG45 SPG65; 5'-nucleotidase (purine), cytosolic type B; Cytosolic 5'-nucleotidase II; Cytosolic purine 5'-nucleotidase; IMP-specific 5'-NT
Gene Aliases: 2010002I23Rik; cN-II; CnII; ENSMUSG00000025039; Gm9751; GMP; NT5B; NT5C2; NT5CP; PNT5; SPG45; SPG65
UniProt ID: (Human) P49902, (Mouse) Q3V1L4
Entrez Gene ID: (Human) 22978, (Mouse) 76952
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.