Novus Biologicals
Manufacturer Code:NBP131404
Catalog # NBP131404
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human NT5C2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-nucleotidase (purine) cytosolic type B 5'-nucleotidase cytosolic II cN-II Cytosolic 5'-nucleotidase II EC 3.1.3 EC 3.1.3.5 GMP NT5BIMP-specific 5'-NT NT5CP PNT5cytosolic purine 5'-nucleotidase purine 5' nucleotidase; 5'-nucleotidase (purine), cytosolic type B; Cytosolic 5'-nucleotidase II; Cytosolic purine 5'-nucleotidase; IMP-specific 5'-NT
Gene Aliases: cN-II; GMP; NT5B; NT5C2; NT5CP; PNT5; SPG45; SPG65
UniProt ID: (Human) P49902
Entrez Gene ID: (Human) 22978
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.