Abnova
Manufacturer Code:PAB30357
Catalog # 89-032-701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human NT5C2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-nucleotidase (purine), cytosolic type B; cN-II; Cytosolic 5'-nucleotidase II; Cytosolic purine 5'-nucleotidase; GMP; IMP-specific 5'-NT; NT5B; PNT5
Gene Aliases: cN-II; GMP; NT5B; NT5C2; NT5CP; PNT5; SPG45; SPG65
UniProt ID: (Human) P49902
Entrez Gene ID: (Human) 22978
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.